Isohexane (mixture of isomers), GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: 2-Methylpentane

287206AEA 118 EUR
287206A
Isohexane (mixture of isomers), GPR RECTAPUR®
Isohexane (mixture of isomers)
Formula: C₆H₁₄
MW: 86.18 g/mol
Boiling Pt: 60
Melting Pt: –154 °C
Density: 0.653 g/cm³ (25 °C)
Flash Pt: −33 °C
Storage Temperature: Ambient
MDL Number: MFCD00009406
CAS Number: 92112-69-1
EINECS: 295-570-2
UN: 1208
ADR: 3 II
REACH: 01-2119484651-34

Order Now

Specification Test Results

n-Hexane Max. 5 %
Evaporation residue Max. 50 ppm
Total S (Sulphur) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR