Fluorescein disodium salt 98.5-100.5% indicator, Fluka™

Supplier: Honeywell Chemicals

28803-100G 28803-1KG 28803-4X1KG
28803-100GEA 64.2 EUR
28803-100G 28803-1KG 28803-4X1KG
Fluorescein disodium salt 98.5-100.5% indicator, Fluka™
Fluorescein disodium salt
Formula: C₂₀H₁₀Na₂O₅
Melting Pt: 300 °C
Density: 1,601 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167039
CAS Number: 518-47-8
EINECS: 208-253-0
Merck Index: 13,04185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated to the dried substance) Min. 98.5 %
Loss on drying (105°C) Max. 10 %
Zinc (Zn) complying
Chloride (Cl) Max. 0.35 %
Sulfate (SO₄) Max. 1.2 %
UV spectrum complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR