Zinc, powder, AnalaR NORMAPUR® 45 µm for analysis

Supplier: VWR Chemicals
Warning

29062.290EA 147 EUR
29062.290 29062.260
Zinc, powder, AnalaR NORMAPUR® 45 µm for analysis
Zinc
Formula: Zn
MDL Number: MFCD00011291
CAS Number: 7440-66-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay min. 98%
Appearance grayish, heavy powder
Arsenic (As) max. 0,0001%
Cadmium (Cd) max. 0,05%
Copper (Cu) max. 0.005%
Iron (Fe) max. 0,005%
Lead (Pb) max. 0,005%
Tin (Sn) max. 0,001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR