Economy scoop
Supplier: Sampling Systems
A304-450 A304-1000 A304-120 A304-1500 A304-2000
A304-450EA 81.1 EUR
A304-450 300-0660 300-0661 300-0662 300-0663
Economy scoop
Scoops
These scoops made from 304 stainless steel are ideal for general industrial use.
  • High polish
  • Light duty construction
  • Round handle
Please note: This scoop is not suitable for GMP applications.
Order Now
SPECIFICATIONS
Environmentally Preferable