Hydrochloric acid 2 mol/l (2 N), AVS TITRINORM Volumetric

Supplier: VWR Chemicals
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

30025.293EA 25.6 EUR
30025.293 30025.362 30025.373 30025.407 30025.320
Hydrochloric acid 2 mol/l (2 N), AVS TITRINORM Volumetric
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Density: 1.031 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 1.996 - 2.004 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR