Potassium iodate 99.7-100.4%, Puriss. p.a., Fluka™

Supplier: Honeywell Chemicals
Danger

30314-100G 30314-1KG 30314-250G 30314-25KG
30314-100GEA 0 EUR
30314-100G 30314-1KG 30314-250G 30314-25KG
Potassium iodate 99.7-100.4%, Puriss. p.a., Fluka™
Potassium iodate
Formula: KlO₃
Melting Pt: 560 °C
Density: 3,93 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1,II
Merck Index: 13,07726

Order Now

Specification Test Results

Assay 99.7 - 100.4 %
Water insoluble matter Max. 0.005 %
Loss on drying (130°C) Max. 0.05 %
pH (5 %, 25 °C) 5.0 - 8.0
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.001 %
Sodium (Na) Max. 0.005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 0.0005 %
Chlorate, Chloride, Bromide, Bromate (as Cl) Max. 0.01 %
Total N Max. 0.002 %
Iodide (I) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR