SuperClear® Microcentrifuge Tubes without Caps
Supplier: LABCON
3033-870-000-9
3033-870-000-9EA 0 EUR
3033-870-000-9
SuperClear® Microcentrifuge Tubes without Caps
Tubes Microcentrifuge Tubes
PP, clear. These microcentrifuge tubes are complete with side graduations and a frosted writing surface.
  • ATP, RNase/DNase and endotoxin-free (non-pyrogenic)
  • Polypropylene resin, USP Class VI
  • Freezable to −80 °C
Packaging: Supplied in resealable bags.