HEPES sodium salt ≥99.5% for biopharmaceutical production

Supplier: VWR Chemicals

30567.297EA 618 EUR
30567.297 30567.366 30567.468
HEPES sodium salt ≥99.5% for biopharmaceutical production
HEPES sodium salt
Formula: C₈H₁₇N₂NaO₄S
Melting Pt: > 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00036463
CAS Number: 75277-39-3
EINECS: 278-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116e9b4f3c17d013ff2b7c9848c219943a725b3c&vid=8c800de68c4434c3c9d77107ff6779ef516bd851&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %
Appearance White crystalline powder
Cell culture toxicity Passes test
DNases Not detected
Identification by IR Conforms to structure
Proteases Not detected
RNases Not detected
Solubility (1 mol/l) Clear and complete
pH (1 %) 9.5 - 10.5
pKa (25°C) 7.4 - 7.7
Heavy metals (as Pb) Max. 5 ppm
Loss on drying (105°C) Max. 2.0 %
Ca (Calcium) Max. 5 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 5 ppm
Mg (Magnesium) Max. 5 ppm
Mn (Manganese) Max. 5 ppm
Zn (Zinc) Max. 5 ppm
Absorbance (260 nm) (1 mol/l) Max. 0.5
Absorbance (280 nm) (1 mol/l) Max. 0.5
Bioburden Max. 100 CFU/g
Endotoxin (0,2 %) Max. 0.1 EU/mg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR