qPCR master mixes, Fast Plus EvaGreen®

Supplier: Biotium

31014 31014-1 31014-2 31015 31015-1 31015-2 31014-T 31015-T 31020-1 31020-2 31020-T 31020
31014.EA 297 EUR
31014. 31014-1 31014-2 31015. 31015-1 31015-2 31014-T 31015-T 31020-1 31020-2 31020-T 31020.
qPCR master mixes, Fast Plus EvaGreen®
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits

A qPCR master mix containing Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase, suitable for qPCR using a fast cycling protocol.


  • 2X qPCR master mix for fast cycling protocols
  • EvaGreen® dye-based
  • Contains Cheetah™ Taq hotstart DNA polymerase


Fast Plus EvaGreen® qPCR Master Mix contains Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase. It is suitable for qPCR using a fast cycling protocol. However for most PCR applications we recommend the Forget-Me-Not™ series of master mixes, for superior activity.


Fast Plus Evagreen® qPCR Master Mix is a 2X master mix, and is supplied with a separate vial of ROX reference dye, for use with high ROX, low ROX, or no ROX instruments.

Order Now

SPECIFICATIONS

Application qPCR dye qPCR dye
Environmentally Preferable Safer Human and Environmental Health Safer Human and Environmental Health
Environmental certification Classified as non-hazardous to aquatic life under CCR title 22 regulations

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR