Potassium hydrogen sulphate ≥99%, Puriss. p.a., Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Potassium bisulphate

31260-50KG 31260-1KG 31260-6X1KG 31260-25KG
31260-50KGEA 2670 EUR
31260-50KG 31260-1KG 31260-6X1KG HONC31260-25KG
Potassium hydrogen sulphate ≥99%, Puriss. p.a., Fluka™
Potassium hydrogen sulphate
Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 200 °C
Density: 2.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8 II
Merck Index: 13,07695

Order Now

Specification Test Results

Assay Min. 99 %
Aluminium (Al) Max. 0.002 %
Arsenic (As) Max. 0.00005 %
Calcium (Ca) Max. 0.002 %
Cadmium (Cd) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Magnesium (Mg) Max. 0.0005 %
Sodium (Na) Max. 0.005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Chloride (Cl) Max. 0.0005 %
Phosphate (PO₄) Max. 0.0005 %
Total N Max. 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR