Potassium iodate 99.4-100.4%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Warning

3156-04EA 160.31 EUR
3156-04
Potassium iodate 99.4-100.4%, BAKER ANALYZED® ACS, J.T.Baker®
Potassium iodate

Lot analysis on label.

Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560
Density: 3.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1 II
Merck Index: 13,07726

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (KIO₃) 99.4 - 100.4 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 5.0 - 8.0
Chloride and Bromide (as Cl) ≤0.01 %
Iodide (I) ≤0.001 %
Nitrogen Compounds (as N) ≤0.005 %
Sulfate (SO₄) ≤0.005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Sodium (Na) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR