Potassium iodide ≥99.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Dipotassium diiodide, Kali iodide

3162-01EA 0 EUR
3162-01 3162-07 3162-09
Potassium iodide ≥99.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330 °C (1013 hPa)
Melting Pt: 686
Density: 3.10 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements ≥99.0%
For Laboratory,Research,or Manufacturing Use ≤0.005%
Assay (KI) ≥99.0 %
Insoluble Matter ≤0.005 %
Loss on Drying at 150°C ≤0.2 %
pH of 5% Solution at 25°C 6.0 - 9.2
Chloride and Bromide (as Cl) ≤0.01 %
Phosphate (PO₄) ≤0.001 %
Sulfate (SO₄) ≤0.005 %
Barium (Ba) ≤0.002 %
Calcium (Ca) ≤0.002 %
Magnesium (Mg) ≤0.001 %
Sodium (Na) ≤0.005 %
Iodate (IO₃) ≤3 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR