VWR® Autoclave Deodorant
Supplier: VWR Chemicals
320201.100EA 196 EUR
320201.100 320301.100 320501.100 320601.100
VWR® Autoclave Deodorant
Deodorants and Air Fresheners
This autoclave deodorant improves the working environment and neutralises odours associated with autoclaving.
  • Capsule releases its contents automatically as the autoclave heats up
  • Easy to use
Order Now
SPECIFICATIONS
Environmentally Preferable