Burettes, lateral stopcock, Schellbach
Supplier: HIRSCHMANN
3210160 3210170 3210175 3210360 3210370 3210375
3210160.EA 61.9 EUR
3210160. 3210170. 3210175. 3210360. 3210370. 3210375.
Burettes, lateral stopcock, Schellbach
Burettes
Burette, class B, DURAN® borosilicate glass 3.3, with blue graduation.
  • Lateral glass stopcock with glass key or PTFE spindle
  • Schellbach stripes for enhanced legibility
Certifications: Calibrated for EX, DIN EN ISO 385.
Order Now
SPECIFICATIONS
Environmentally Preferable