Toluene ≥99.7% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

32249-1L 32249-2.5L 32249-25L 32249-4X2.5L 32249-6X1L 32249-7L
32249-1LEA 60.9 EUR
32249-1L 32249-2.5L 32249-25L 32249-4X2.5L 32249-6X1L 32249-7L
Toluene ≥99.7% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
Toluene

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 635760

Formula: C₆H₅CH₃
Boiling Pt: 110,6 °C
Melting Pt: -95 °C
Density: 0,867 g/cm³ (20 °C)
Flash Pt: 7,1 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Pressure 26 mmHg ( 25 °C)
Assay (GC) Min. 99.7 %
Density (D 20/20) 0.865 - 0.869
Refractive index (n 20/D) 1.496 - 1.498
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.02 %
Free acid (as HCl) Max. 0.001 %
Free alkali (as NaOH) Max. 0.002 %
Aluminium (Al) Max. 0.00005 %
Barium (Ba) Max. 0.00001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Potassium (K) Max. 0.00005 %
Lithium (Li) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Molybdenum (Mo) Max. 0.00001 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Strontium (Sr) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
S-compounds (as S) Max. 0.003 %
Thiophene Max. 0.0001 %
Reaction against H₂SO₄ complying
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR