Ammonium acetate ≥98%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Chemicals

32301-1KG 32301-2.5KG 32301-500G 32301-6X1KG 32301-100G 32301-10KG 32301-20KG 32301-10KG-SE
32301-1KGEA 133 EUR
32301-1KG 32301-2.5KG 32301-500G 32301-6X1KG 32301-100G 32301-10KG 32301-20KG HONC32301-10KG-SE
Ammonium acetate ≥98%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Ammonium acetate
Formula: H₃CCOONH₄
Melting Pt: 113 °C
Density: 1,170 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %
Water insoluble matter Max. 0.005 %
Water (Karl Fischer) Max. 2 %
Sulfated ash Max. 0.01 %
pH (5 %, 25 °C) 6.7 - 7.3
Calcium (Ca) Max. 0.001 %
Cadmium (Cd) Max. 0.0002 %
Cobalt (Co) Max. 0.0005 %
Copper (Cu) Max. 0.0002 %
Chromium (Cr) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.0002 %
Manganese (Mn) Max. 0.0005 %
Sodium (Na) Max. 0.005 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.0002 %
Zinc (Zn) Max. 0.0002 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.0005 %
Nitrate (NO₃) Max. 0.001 %
Sulfate (SO₄) Max. 0.001 %
KMnO₄ reducing matter (as HCOOH) Max. 0.005 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR