Acetonitrile, extra dry 99.9+%, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

326811000 326810010 326810025
326811000.EA 97.8 EUR
326811000. 326810010. 326810025.
Acetonitrile, extra dry 99.9+%, AcroSeal®
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81…82 °C (760 mmHg)
Melting Pt: –46 °C
Density: 0,781 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.001 % (Coulometric)
GC ≥99.90 %
Residue after evaporation ≤0.0005 %
Acidity (CH3COOH) ≤0.002 %
Acetamide ≤10 ppm

SPECIFICATIONS

Environmentally Preferable Safer Human and Environmental Health
Environmental certification EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR