Bromophenol blue Reag. Ph. Eur. Indicator, Fluka™

Supplier: Honeywell Chemicals
Warning

Synonyms: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein

32712-5G 32712-25G
32712-5GEA 53.5 EUR
32712-5G 32712-25G
Bromophenol blue Reag. Ph. Eur. Indicator, Fluka™
Bromophenol blue
Formula: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
Melting Pt: 204 °C
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2
Merck Index: 13,01429

Order Now

Specification Test Results

Indicator solution complying
Test of sensitivity complying
Change range (pH 2,8-4,4) complying
UV spectrum complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR