Rods, ceramic, round, solid, AL23
Supplier: Thermo Fisher Scientific
32921.DM 32931.DR 32931.DT 32931.DW 32931.EH 32933.DR 32933.DT 32933.DW 32933.EM
32921.DMEA 0 EUR
32921.DM 32931.DR 32931.DT 32931.DW 32931.EH 32933.DR 32933.DT 32933.DW 32933.EM
Rods, ceramic, round, solid, AL23
Solid Rods
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Order Now
SPECIFICATIONS
Environmentally Preferable