VWR®, Jerricans, Wide Neck, HDPE, with Screw Caps
Supplier: VWR Collection
331-0152EA 6.5 EUR
331-0152 331-0187 331-0103 331-0107 331-0108 331-0112 331-0411 VWRI331-0109.1
VWR®, Jerricans, Wide Neck, HDPE, with Screw Caps
Bottles Jerricans
HDPE, opaque with PP screw caps.
  • Space saving container, easy to fill
  • With tamper-evident screw caps
  • Excellent shock resistance
  • Temperature resistance: −10 to +80 °C (120 °C for short periods)
  • Can be supplied sterile
Ideal for water and general testing, filling and storage. Opaque, ensures reasonable visibility of the content level.