Micro burettes, bang pattern, lateral stopcock, Schellbach
Supplier: HIRSCHMANN
3310140 3310152 3310160 3310340 3310352 3310360
3310140.EA 192 EUR
3310140. 3310152. 3310160. 3310340. 3310352. 3310360.
Micro burettes, bang pattern, lateral stopcock, Schellbach
Burettes
Micro burette, class AS, DURAN®, blue graduation, certified conformity, with dated batch identification.
  • Lateral stopcock with standard ground glass key or PTFE spindle, with socket, Bang
  • Schellbach stripes for enhanced legibility
  • Main point ring graduation
Certifications: Calibrated for EX, DIN EN ISO 385.