tert-Butanol 99+% ACS

Supplier: Thermo Fisher Scientific
Danger

33278.AK 33278.K2 33278.K7 33278.M1
33278.AKEA 35.8 EUR
33278.AK 33278.K2 33278.K7 33278.M1
tert-Butanol 99+% ACS
tert-Butanol

Purity: 99+% CAS No.: 75-65-0 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: 11°(52°F) Physical Form: Liquid Density: 0.786 Refractive Index: 1.3870 Appearance: Colourless Melting Point: 25° Boiling Point: 83° Beilstein Reference: 906698 Merck Reference: 14,1542 UN No.: 1120 MDL No.: MFCD00004464

Formula: (CH₃)₃COH
Boiling Pt: 83 °C
Melting Pt: 25 °C
Density: 0,786
Flash Pt: 11 °C (52 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3,II
Merck Index: 14,1542
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a1b0c51eec643e5d5b7152a3e410db93d1626e&vid=999328a3c31895b190d09313b24501682921af01&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR