Copper(II) sulphate, anhydrous, reagent

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Cupric sulphate

33308.22 33308.36 33308.A3
33308.22EA 58.4 EUR
33308.22 33308.36 33308.A3
Copper(II) sulphate, anhydrous, reagent
Copper (II) sulphate

CAS No.: 7758-98-7 Molecular Formula: CuO4S Formula Weight: 159.60 Physical Form: Powder Density: 3.6 Sensitivity: Hygroscopic Appearance: Pale grey Odour: Odourless Melting Point: 560° dec. Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,2653 UN No.: 3288 MDL No.: MFCD00010981

Formula: CuSO₄
MW: 159.61 g/mol
Melting Pt: >560 °C (dec.)
Density: 3.6
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3288
ADR: 6.1 III
Merck Index: 14,2653

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR