Magnesium hydroxide carbonate tetrahydrate (∼40 - 43,5% MgO), Reagent Grade

Supplier: Thermo Fisher Scientific

33333.36 33333.22
33333.36EA 97.6 EUR
33333.36 33333.22
Magnesium hydroxide carbonate tetrahydrate (∼40 - 43,5% MgO), Reagent Grade
Magnesium hydroxide carbonate tetrahydrate

CAS No.: 39409-82-0 Molecular Formula: 4MgCO3.Mg(OH)2.4H2O approx. Formula Weight: 467.67 (395.58anhy) Physical Form: -325 Mesh Powder Appearance: White Odour: Odourless Merck Reference: 14,5660 MDL No.: MFCD00149783

Formula: 4MgCO₃·Mg(OH)₂·4H₂O
Boiling Pt: 3600 °C (1013 hPa)
Melting Pt: > 2500 °C
Density: 2,16 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149783
CAS Number: 235-192-7
EINECS: 235-192-7
Merck Index: 14,5660

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR