(S)-(+)-2-Heptanol ≥98%

Supplier: Thermo Fisher Scientific
Warning

33795.03
33795.03EA 0 EUR
33795.03
(S)-(+)-2-Heptanol ≥98%
(S)-(+)-2-Heptanol

Purity: 98% CAS No.: 6033-23-4 Molecular Formula: C7H16O Formula Weight: 116.20 Flash Point: 59°(138°F) Physical Form: Liquid Density: 0.815 Refractive Index: 1.4210 Boiling Point: 160-162° Merck Reference: 14,4662 UN No.: 1987 MDL No.: MFCD00065003

Formula: C₇H₁₆O
Boiling Pt: 160…162 °C
Density: 0,815
Flash Pt: 59 °C (138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00065003
CAS Number: 6033-23-4
UN: 1987
ADR: 3,III
Merck Index: 14,4662

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR