Ammonium chloride ≥99.5% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning


Management of Change (MOC) category = N

3384-12EA 0 EUR
3384-12 3384-09 3384-20
Ammonium chloride ≥99.5% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™
Ammonium chloride
Formula: NH₄Cl
Melting Pt: 335 °C
Density: 1,53 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements ≥99.5%
For Laboratory,Research,or Manufacturing Use ≤0.001%
ACS - Assay (NH₄Cl) (by Ag titrn) ≥99.5 %
ACS - Calcium (Ca) ≤0.001 %
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤2 ppm
ACS - Magnesium (Mg) ≤5 ppm
ACS - pH of 5% Solution at 25°C 4.5 - 5.5
ACS - Phosphate (PO₄) ≤2 ppm
ACS - Residue after Ignition ≤0.01 %
ACS - Sulfate (SO₄) ≤0.002 %
ACS - Appearance (white crystals) Passes Test
Heavy Metals (as Pb) ≤2 ppm
Nitrate (NO₃) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR