Xylene (mixture of isomers), J.T.Baker®

Supplier: Avantor
Danger

3410.2500PEEA 0 EUR
3410.2500PE 3410.5000PE 3410.9010 3410.9025
Xylene (mixture of isomers), J.T.Baker®
Xylene (mixture of isomers)
Formula: C₆H₄(CH₃)₂
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –34 °C
Density: 0,86…0.88 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Benzene max. 0.02 %
Color (APHA) max. 20
Density (g/ml) at 20°C 0.862 - 0.872

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR