Perchloric acid 0.01 M HClO4 (0.01N) acetic acid Volumetric, Titration_nonaqueous_liquids, Fluka™

Supplier: Honeywell Chemicals
Danger

34408-1L
34408-1LEA 480 EUR
34408-1L
Perchloric acid 0.01 M HClO4 (0.01N) acetic acid Volumetric, Titration_nonaqueous_liquids, Fluka™
Perchloric acid
Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 119 °C (1.013 hPa) (solvent)
Melting Pt: -101
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 40 °C (solvent)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
UN: 2920
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

c(perchloric acid) 9.95 - 10.05 mM
Water (Karl Fischer) 0.05 - 0.15 %
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR