Ethanol absolute (taxed) ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

34852-7L 34852-2.5L 34852-1L 34852-4X2.5L
34852-7LEA 2110 EUR
34852-7L 34852-2.5L 34852-1L 34852-4X2.5L
Ethanol absolute (taxed) ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Ethanol

Caution: Ethanol (Taxed) for domestic sale in Germany, France, Ireland and United Kingdom

Formula: C₂H₅OH
Boiling Pt: 78…79 °C (1,013 hPa)
Melting Pt: –115 °C
Density: 0,790…0.791 g/cm3 (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now

Specification Test Results

Assay (GC) Min. 99.8 Vol.%
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.2 %
Free acid (as CH₃COOH) Max. 0.001 %
Transmittance at 210 nm Min. 20 %
Transmittance at 225 nm Min. 50 %
Transmittance at 240 nm Min. 80 %
Transmittance from 260 nm Min. 98 %
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
Autoignition Temperature 683 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR