Cumene hydroperoxide 80% in aqueous solution

Supplier: Thermo Fisher Scientific
Danger

349960050 349962500 349960010 349960025
349960050.EA 28.2 EUR
349960050. 349962500. 349960010. 349960025.
Cumene hydroperoxide 80% in aqueous solution
Cumene hydroperoxide
Formula: C₉H₁₂O₂
Melting Pt: –30 °C
Density: 1,06 g/cm³
Flash Pt: 83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002129
CAS Number: 80-15-9
EINECS: 201-254-7
UN: 3109
ADR: 5.2,

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Titration Iodometric 80 to 85 %
Color scale ≤450 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR