Automatic burettes, pellet pattern, lateral stopcock and intermediate valve
Supplier: HIRSCHMANN
352016060 352017060 352017560 352036060 352037060 352037560
352016060.EA 291 EUR
352016060. 352017060. 352017560. 352036060. 352037060. 352037560.
Automatic burettes, pellet pattern, lateral stopcock and intermediate valve
Burettes
Automatic burette, Pellet pattern, class B, brown glass, DURAN® with white graduation.
  • Lateral PTFE stopcock or glass key
  • With intermediate valve
Certifications: Compliance with DIN EN ISO 385.