Ammonium sulphate 99,0-100,5, granular NF, ACS, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

3523-65EA 0 EUR
3523-65
Ammonium sulphate 99,0-100,5, granular NF, ACS, J.T.Baker®
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Density: 1.77 g/cm³
MDL Number: MFCD00003391
CAS Number: 7783-20-2
Merck Index: 13,00559

Order Now

Specification Test Results

Meets N.F. Requirements
GMP Manufactured Product
Meets ACS Reagent Chemical Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 99.0 - 100.5 %
NF - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
NF - Insoluble Matter ≤0.005 %
NF - Residue on Ignition ≤0.005 %
NF - pH of 5% Solution 5.0 - 6.0
NF - Nitrate (NO₃) ≤0.001 %
NF - Chloride (Cl) ≤5 ppm
NF - Phosphate (PO₄) ≤5 ppm
NF - Iron (Fe) ≤5 ppm
NF - Identification Passes Test
NF - Total Aerobic Microbial Count ≤100 cfu/g
NF - Total Yeast and Mold Count ≤10 cfu/g
ACS - Assay ((NH₄)₂SO₄) (A.C.S.) ≥99.0 %
ACS - Residue on Ignition ≤0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 6.0
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤3 ppm
ACS - Nitrate (NO₃) ≤0.001 %
ACS - Phosphate (PO₄) ≤3 ppm
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Iron (Fe) ≤5 ppm
Appearance (White crystals or granules) Passes Test
No Class 1, 2 or 3 Residual Solvents are likely to be present by
exclusion of the production/purification process.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.
Preserve in well closed container.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR