Hydrochloric acid 0.05 M (0.05 N) volumetric solution, Fluka™

Supplier: Honeywell Chemicals
Warning

35320-1L 35320-6X1L
35320-1LEA 37.3 EUR
35320-1L 35320-6X1L
Hydrochloric acid 0.05 M (0.05 N) volumetric solution, Fluka™
Hydrochloric acid
Formula: HCl
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1,0 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-791-2
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Vapor Pressure 190 mmHg ( 25 °C)
Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR