Barium chloride 20% in aqueous solution, Fluka™

Supplier: Honeywell Chemicals
Warning

35323-5L
35323-5LEA 236 EUR
35323-5L
Barium chloride 20% in aqueous solution, Fluka™
Barium chloride
Formula: BaCl₂
MW: 208.24 g/mol
Boiling Pt: 1560
Melting Pt: 963
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
Merck Index: 12,00998

Order Now

Specification Test Results

Assay 19 - 21 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR