Hydrochloric acid 4.0 M HCl (4N) Volumetric, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

35402-10L
35402-10LEA 423 EUR
35402-10L
Hydrochloric acid 4.0 M HCl (4N) Volumetric, Fluka™
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Density: 1.072 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Vapor Pressure 190 mmHg ( 25 °C)
Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR