2,6-Dichloroquinone-4-chloroimide, Supelco®

Supplier: Merck


M-Clarity™ quality level = MQ100

35620-10G 35620-50G
35620-10GEA 121 EUR
35620-10G 35620-50G
2,6-Dichloroquinone-4-chloroimide, Supelco®
2,6-Dichloroquinone-4-chloroimide
Formula: C₆H₂Cl₃NO
MDL Number: MFCD00001611
CAS Number: 101-38-2
EINECS: 202-937-2
UN: 3224
ADR: 4.1,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ed958fa04c0c787b52af31a24b175a2935c2175&vid=997d36357d204d93163fe875e0cb1fb64f62f360&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR