Erbium triiodide, ultra dry ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Fisher Scientific


ampouled under argon

35695.03 35695.06
35695.03EA 120 EUR
35695.03 35695.06
Erbium triiodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Erbium triiodide

Purity: 99.9% CAS No.: 13813-42-8 Molecular Formula: ErI3 Formula Weight: 547.97 Physical Form: Pieces/Powder, Ampouled under argon Sensitivity: Hygroscopic Boiling Point: 1280° MDL No.: MFCD00016075

Formula: ErI₃
Boiling Pt: 1280 °C
Melting Pt: 1020 °C
Density: ∼5,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016075
CAS Number: 13813-42-8
EINECS: 237-471-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR