Strontium chloride, ultra dry ≥99.995% (metals basis)

Supplier: Thermo Fisher Scientific
Danger


ampouled under argon

35719.06 35719.14
35719.06EA 126 EUR
35719.06 35719.14
Strontium chloride, ultra dry ≥99.995% (metals basis)
Strontium chloride

Purity: 99.995% CAS No.: 10476-85-4 Molecular Formula: SrCl2 Formula Weight: 158.53 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.052 Refractive Index: 1.650 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 873° Boiling Point: 1250° Merck Reference: 14,8840 MDL No.: MFCD00011249

Formula: Cl₂Sr
Boiling Pt: 1250 °C
Melting Pt: 873 °C
Density: 3,052
Storage Temperature: Ambient
MDL Number: MFCD00011249
CAS Number: 10476-85-4
EINECS: 233-971-6
Merck Index: 14,8840

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR