Iridium standard solution, 1000 mg/L Ir in 20% hydrochloric acid (from IrCl₃), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

35751.AD 35751.AE 35751.AP
35751.ADEA 127 EUR
35751.AD 35751.AE 35751.AP
Iridium standard solution, 1000 mg/L Ir in 20% hydrochloric acid (from IrCl₃), standard for ICP, Specpure®
Standards Cationic Standards Iridium Standards

Molecular Formula: IrCl₃ in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011062


Certifications: Traceable to NIST

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00011062

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR