Gas cylinder holder
Supplier: R L ENTERPRISES
3603
3603.EA 154 EUR
3603.
Gas cylinder holder
Wall Hooks
This gas cylinder holder is made of heavy duty alloy casting that is secured to the benches up to 70 mm thick with two plated steel tommy screws. Two countersunk holes are provided to accept wood screws for permanent fixing.
  • Suitable for gas cylinders up to 300 mm diameter
  • Complete with fire resistant belt and buckle