Crucibles
Supplier: Thermo Fisher Scientific
35886.KT 35928.KT 35932.KT 35953.KT 35994.KT 36012.KT 36016.KT 36037.KT 36056.KT 36072.KT 36085.KT 36090.KT 36099.KT
35886.KTEA 107 EUR
35886.KT 35928.KT 35932.KT 35953.KT 35994.KT 36012.KT 36016.KT 36037.KT 36056.KT 36072.KT 36085.KT 36090.KT 36099.KT
Crucibles
Crucibles
These crucibles are manufactured from Inconel® nickel-chromium-iron alloy 601. Available in low form and straight wall types.
  • Excellent resistance to oxidation in the 1000 to 1200 °C temperature range
  • Good corrosion resistance to many acid and aqueous salt solutions
  • Unaffected by rapid changes from hot to cold
  • Retains its mechanical strength at elevated temperatures
Designed for applications that require resistance to heat and corrosion. The high resistance of Inconel® alloy 601 to oxidation, carburisation or sulfidation make it well suited forMore Product Information