Paraffin wax, Solidification point: 55 - 58 °C, pastille, Gurr™

Supplier: VWR Chemicals

Synonyms: Wax Paraffin, Paraffin wax

361077EEA 187 EUR
361077E
Paraffin wax, Solidification point: 55 - 58 °C, pastille, Gurr™
Paraffin
Density: 0.9 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
CAS Number: 8002-74-2

Order Now

Specification Test Results

Appearance (65 °C) Passes test
Congealing range 55 - 58 °C
Cutting/ ribboning Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR