Aluminium sulphate hydrate 98.0-102% ACS

Supplier: Thermo Fisher Scientific
Danger

36294.30 36294.A1
36294.30EA 54.6 EUR
36294.30 36294.A1
Aluminium sulphate hydrate 98.0-102% ACS
Aluminium sulphate hydrate

Purity: 98.0-102% CAS No.: 17927-65-0 Molecular Formula: Al2(SO4)3.xH2O (x≈14-18) Formula Weight: 342.15(anhy) Physical Form: Crystalline Density: 1.61 pH: 3.5 Merck Reference: 14,366 MDL No.: MFCD00149136

Formula: Al₂(SO₄)₃·xH₂O
Density: 1,61
Storage Temperature: Ambient
MDL Number: MFCD00149136
CAS Number: 17927-65-0
EINECS: 233-135-0
Merck Index: 14,366

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR