N-Methyl-2-pyrrolidone (NMP), extra dry over Molecular Sieve 99.5%, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

364381000 364380010 364380025 364385000
364381000.EA 44.9 EUR
364381000. 364380010. 364380025. 364385000.
N-Methyl-2-pyrrolidone (NMP), extra dry over Molecular Sieve 99.5%, AcroSeal®
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
Boiling Pt: 202 °C (760 mmHg)
Melting Pt: –24 °C
Density: 1,03 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid (May darken on storage)
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.5 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR