Copper 99,9 Metals basis, shots ACS ≤ 6 mm (0.2 in)

Supplier: Thermo Fisher Scientific

36482.22 36482.36
36482.22EA 57.9 EUR
36482.22 36482.36
Copper 99,9 Metals basis, shots ACS ≤ 6 mm (0.2 in)
Copper

Purity: 99.9+% CAS No.: 7440-50-8 Odour: Odourless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR