Dysprosium(III) isopropoxide 5% (w/v) in 2-propanol/toluene , 99.9% (REO, rare earth oxide basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Dysprosium(III) i-propoxide

36585.AA 36585.AC 36585.AE
36585.AAEA 0 EUR
36585.AA 36585.AC 36585.AE
Dysprosium(III) isopropoxide 5% (w/v) in 2-propanol/toluene , 99.9% (REO, rare earth oxide basis)
Dysprosium (III) isopropoxide

Purity: 99.9% CAS No.: 6742-68-3 Molecular Formula: C9H21DyO3 Formula Weight: 339.76 Physical Form: Liquid Sensitivity: Moisture Sensitive Appearance: Brown UN No.: 1993 MDL No.: MFCD00049784

Formula: C₉H₂₁DyO₃
Storage Temperature: Ambient
MDL Number: MFCD00049784
CAS Number: 6742-68-3
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR