Palladium 3% on activated carbon wetted with ca. 50% water, powder hydrogenation catalyst, reduced

Supplier: Thermo Fisher Scientific

38296.06 38296.14 38296.22
38296.06EA 66.8 EUR
38296.06 38296.14 38296.22
Palladium 3% on activated carbon wetted with ca. 50% water, powder hydrogenation catalyst, reduced
Palladium

CAS No.: 7440-05-3 Physical Form: 900-1100m2/g Odour: Odourless MDL No.: MFCD03457879

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR