Dry ice storage tubes in IntegraPack®, ProtectR®
Supplier: LABCON
3830-320-000-9 3830-325-000-9 3880-320-000-9 3880-325-000-9
3830-320-000-9EA 151 EUR
3830-320-000-9 3830-325-000-9 3880-320-000-9 3880-325-000-9
Dry ice storage tubes in IntegraPack®, ProtectR®
Tubes Centrifuge Tubes
ProtectR® tube is the only container closure system engineered to be CO₂ resistant and stop costly damage to molecular and cellular solutions when using dry ice. Made of durable, high-quality materials and fitted with an air-tight cap, ProtectR® Tubes form a protective barrier impermeable to CO₂, keeping the sample pH stable and preventing acidification.
  • ATP-, RNase/DNase-, and endotoxin-free (non-pyrogenic)
  • Attached CO₂ resistant flat caps, certified to prevent acidification from dry ice shipment or storage
  • Caps feature CO₂ resistant elastomeric seal
More Product Information