Palladium 5% on activated carbon paste for use as a catalyst A102023-5

Supplier: Thermo Fisher Scientific

38300.06 38300.14 38300.22
38300.06EA 95.5 EUR
38300.06 38300.14 38300.22
Palladium 5% on activated carbon paste for use as a catalyst A102023-5
Palladium

Appearance: Black Odor: Odorless


Ordering information: Sold on a dry weight basis. Unit weight excludes water weight. Nominally 50% water wet

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR