Palladium 10% on activated carbon powder wetted with ca. 50% water for use as a catalyst, unreduced

Supplier: Thermo Fisher Scientific

38305.06 38305.14 38305.22
38305.06EA 191 EUR
38305.06 38305.14 38305.22
Palladium 10% on activated carbon powder wetted with ca. 50% water for use as a catalyst, unreduced
Palladium

Physical Form: 900-1100 m²/g

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR